Mesitylessigsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD13255-5GEA 6 EUR
BLDPBD13255-5G BLDPBD13255-10G BLDPBD13255-100G BLDPBD13255-25G
Mesitylessigsäure 97%
Mesitylessigsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014368
CAS-Nummer: 4408-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR