Ethyl α-chlorphenylacetat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD129255-5GEA 41 EUR
BLDPBD129255-5G BLDPBD129255-10G BLDPBD129255-1G
Ethyl α-chlorphenylacetat 97%
Ethyl α-chlorphenylacetat

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁ClO₂
Molecular Weight: 198,6461 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00013675
CAS-Nummer: 4773-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR