2',4'-Dimethoxy-2,2,2-trifluoracetophenon 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD12810-5GEA 1046 EUR
BLDPBD12810-5G BLDPBD12810-1G BLDPBD12810-100MG BLDPBD12810-250MG
2',4'-Dimethoxy-2,2,2-trifluoracetophenon 97%
2',4'-Dimethoxy-2,2,2-trifluoracetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉F₃O₃
Molecular Weight: 234,1719 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00835330
CAS-Nummer: 578-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR