Ethylthiazol-5-carboxylat 98%

BLDPBD127844-250MGEA 7 EUR
BLDPBD127844-250MG BLDPBD127844-5G BLDPBD127844-10G BLDPBD127844-1G BLDPBD127844-25G
Ethylthiazol-5-carboxylat 98%
Ethylthiazol-5-carboxylat

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₇NO₂S
Molecular Weight: 157,1900 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06205069
CAS-Nummer: 32955-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR