Enalaprilat dihydrate 97%

BLDPBD127253-1GEA 325 EUR
BLDPBD127253-1G BLDPBD127253-100MG BLDPBD127253-50MG BLDPBD127253-250MG
Enalaprilat dihydrate 97%
Enalaprilat dihydrate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₈H₂₄N₂O₅·2H₂O
Molecular Weight: 348,40 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00941393
CAS-Nummer: 84680-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR