Doripenem monohydrate 98%

BLDPBD126854-1GEA 122 EUR
BLDPBD126854-1G BLDPBD126854-100MG BLDPBD126854-250MG BLDPBD126854-5G
Doripenem monohydrate 98%
Doripenem monohydrate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₄N₄O₆S₂·H₂O
Molecular Weight: 420,50 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09749887
CAS-Nummer: 364622-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR