D(-)-Luciferin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD126657-100MGEA 66 EUR
BLDPBD126657-100MG
D(-)-Luciferin 98%
D(-)-Luciferin

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₁H₈N₂O₃S₂
Molecular Weight: 280,3228 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00042929
CAS-Nummer: 2591-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR