Dimethyl-cis-cyclohexan-1,2-dicarboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD125391-500GEA 1609 EUR
BLDPBD125391-500G BLDPBD125391-10G BLDPBD125391-5G BLDPBD125391-1G BLDPBD125391-100G BLDPBD125391-25G
Dimethyl-cis-cyclohexan-1,2-dicarboxylat 97%
Dimethyl-cis-cyclohexan-1,2-dicarboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₆O₄
Molecular Weight: 200,2300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043768
CAS-Nummer: 1687-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR