Diethylphenylphosphin 98+%
|
|
BLDPBD124904-250MGEA
17
EUR
BLDPBD124904-250MG
BLDPBD124904-5G
BLDPBD124904-1G
Diethylphenylphosphin 98+%
Diethylphenylphosphin
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₀H₁₅P Molecular Weight: 166,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015172 CAS-Nummer: 1605-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...