4-tert-Butylphenylhydrazin Hydrochlorid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD12321-500GEA 1444 EUR
BLDPBD12321-500G BLDPBD12321-0.5G BLDPBD12321-1G BLDPBD12321-250MG BLDPBD12321-10G BLDPBD12321-25G BLDPBD12321-5G BLDPBD12321-100G
4-tert-Butylphenylhydrazin Hydrochlorid 95%
4-tert-Butylphenylhydrazin Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₇ClN₂
Molecular Weight: 200,7084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013387
CAS-Nummer: 128231-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR