Cyclohexyl(dimethoxy)methylsilan 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD122797-25GEA 7 EUR
BLDPBD122797-25G
Cyclohexyl(dimethoxy)methylsilan 98%
Cyclohexyl(dimethoxy)methylsilane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₂₀O₂Si
Molecular Weight: 188,3394 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00071752
CAS-Nummer: 17865-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR