3-Thiophenboronsäurepinakolester 96%
|
|
BLDPBD12255-1GEA
25
EUR
BLDPBD12255-1G
BLDPBD12255-5G
BLDPBD12255-10G
BLDPBD12255-250MG
BLDPBD12255-25G
3-Thiophenboronsäurepinakolester 96%
3-Thiophenboronsäurepinakolester
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₁₀H₁₅BO₂S Molecular Weight: 210,1009 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD05663893 CAS-Nummer: 214360-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...