Kupfer(II)sulfat Pentahydrat 99%

Synonyme: Kupfersulfat-PentahydratKupfersulfat Pentahydrat

BLDPBD122189-25GEA 7 EUR
BLDPBD122189-25G
Kupfer(II)sulfat Pentahydrat 99%
Kupfer(II)sulfat Pentahydrat

Storage condition: Inert atmosphere, Room Temperature

Formel: CuSO₄·5H₂O
Molecular Weight: 249,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149681
CAS-Nummer: 7758-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR