Methyl-(3-methoxyphenyl)acetat 98%

BLDPBD12057-5GEA 19 EUR
BLDPBD12057-5G BLDPBD12057-0.025G BLDPBD12057-10G BLDPBD12057-100G BLDPBD12057-1G BLDPBD12057-25G
Methyl-(3-methoxyphenyl)acetat 98%
Methyl-(3-methoxyphenyl)acetat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₂O₃
Molecular Weight: 180,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017205
CAS-Nummer: 18927-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR