(1-Hexadecyl)tributylphosphoniumbromid 97%
|
|
BLDPBD120539-100GEA
165
EUR
BLDPBD120539-100G
BLDPBD120539-500G
BLDPBD120539-5G
BLDPBD120539-10G
BLDPBD120539-25G
(1-Hexadecyl)tributylphosphoniumbromid 97%
(1-Hexadecyl)tributylphosphoniumbromid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₂₈H₆₀BrP Molecular Weight: 507,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011775 CAS-Nummer: 14937-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...