Picamilon 98%

BLDPBD11992-25GEA 221 EUR
BLDPBD11992-25G BLDPBD11992-250MG BLDPBD11992-10G BLDPBD11992-5G BLDPBD11992-1G
Picamilon 98%
Picamilon

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00186518
CAS-Nummer: 34562-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR