Calciumchlorid 96%

Synonyme: Chlorcalcium

BLDPBD119632-1000GEA 10 EUR
BLDPBD119632-1000G BLDPBD119632-100G BLDPBD119632-500G
Calciumchlorid 96%
Calciumchlorid

Storage condition: Sealed in dry, Room Temperature

Formel: CaCl₂
Molecular Weight: 110,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010903
CAS-Nummer: 10043-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR