2-Isopropoxy-5-(trifluormethyl)phenylboronsäure 98%

BLDPBD11946-5GEA 410 EUR
BLDPBD11946-5G BLDPBD11946-250MG BLDPBD11946-1G
2-Isopropoxy-5-(trifluormethyl)phenylboronsäure 98%
2-Isopropoxy-5-(trifluormethyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₂BF₃O₃
Molecular Weight: 248,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07363786
CAS-Nummer: 850593-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR