Methyl-4-acetylbenzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11842-25GEA 48 EUR
BLDPBD11842-25G BLDPBD11842-100G BLDPBD11842-5G BLDPBD11842-500G BLDPBD11842-10G
Methyl-4-acetylbenzoat 98%
Methyl-4-acetylbenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216474
CAS-Nummer: 3609-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR