Benzyldimethyltetradecylammoniumchlorid 98% + (contains < 10% H₂O)
|
|
BLDPBD116285-25GEA
7
EUR
BLDPBD116285-25G
Benzyldimethyltetradecylammoniumchlorid 98% + (contains < 10% H₂O)
Benzyldimethyltetradecylammoniumchlorid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₂₃H₄₂ClN Molecular Weight: 368,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011771 CAS-Nummer: 139-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...