(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 97%

BLDPBD115807-5GEA 213 EUR
BLDPBD115807-5G BLDPBD115807-250MG BLDPBD115807-25G BLDPBD115807-100MG BLDPBD115807-1G
(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 97%
(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₈H₃₈O₅
Molecular Weight: 454,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21603881
CAS-Nummer: 133676-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR