Benzo[b]naphtho[1,2-d]thiophene 97%

BLDPBD115657-100MGEA 22 EUR
BLDPBD115657-100MG BLDPBD115657-1G BLDPBD115657-25G BLDPBD115657-10G BLDPBD115657-250MG BLDPBD115657-5G
Benzo[b]naphtho[1,2-d]thiophene 97%
Benzo[b]naphtho[1,2-d]thiophene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₆H₁₀S
Molecular Weight: 234,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00215942
CAS-Nummer: 205-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR