α,α-Dimethoxytoluol 97%

Synonyme: Benzaldehyddimethylacetal

BLDPBD115374-500GEA 57 EUR
BLDPBD115374-500G BLDPBD115374-1000G BLDPBD115374-100G BLDPBD115374-25G
α,α-Dimethoxytoluol 97%
α,α-Dimethoxytoluol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008491
CAS-Nummer: 1125-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR