Ethyl-2,3,4,5-tetrafluorbenzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11514-5GEA 18 EUR
BLDPBD11514-5G BLDPBD11514-25G BLDPBD11514-100G
Ethyl-2,3,4,5-tetrafluorbenzoat 98%
Ethyl-2,3,4,5-tetrafluorbenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094158
CAS-Nummer: 122894-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR