5-Fluoruridin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11282-25GEA 166 EUR
BLDPBD11282-25G BLDPBD11282-250MG BLDPBD11282-5G BLDPBD11282-10G BLDPBD11282-1G
5-Fluoruridin 97%
5-Fluoruridin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₁FN₂O₆
Molecular Weight: 262,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036832
CAS-Nummer: 316-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR