2-Methoxy-5-(trifluormethyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11096-5GEA 150 EUR
BLDPBD11096-5G BLDPBD11096-1G BLDPBD11096-25G
2-Methoxy-5-(trifluormethyl)phenylboronsäure 98%
2-Methoxy-5-(trifluormethyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈BF₃O₃
Molecular Weight: 219,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039222
CAS-Nummer: 240139-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR