2,4-Dichlor-3-ethyl-6-nitrophenol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11026-25GEA 29 EUR
BLDPBD11026-25G BLDPBD11026-5G BLDPBD11026-10G BLDPBD11026-0.025G
2,4-Dichlor-3-ethyl-6-nitrophenol 98%
2,4-Dichlor-3-ethyl-6-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇Cl₂NO₃
Molecular Weight: 236,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270764
CAS-Nummer: 99817-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR