2,2'-Bithiophen-5-boronsäure 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD11000-5GEA 1086 EUR
BLDPBD11000-5G BLDPBD11000-250MG BLDPBD11000-1G BLDPBD11000-100MG
2,2'-Bithiophen-5-boronsäure 96%
2,2'-Bithiophen-5-boronsäure

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₇BO₂S₂
Molecular Weight: 210,0810 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03086112
CAS-Nummer: 132898-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR