6-Methylimidazo[1,2-a]pyridin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD109742-250MGEA 37 EUR
BLDPBD109742-250MG BLDPBD109742-1G BLDPBD109742-5G BLDPBD109742-100MG
6-Methylimidazo[1,2-a]pyridin 98%
6-Methylimidazo[1,2-a]pyridin

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂
Molecular Weight: 132,1625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663806
CAS-Nummer: 874-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR