6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD108933-500GEA 340 EUR
BLDPBD108933-500G BLDPBD108933-25G BLDPBD108933-5G BLDPBD108933-100G
6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid 98%
6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅Cl₂N₃
Molecular Weight: 190,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258637
CAS-Nummer: 13493-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR