6-Brom-3-nitro-4-chinolinol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD108453-1GEA 8 EUR
BLDPBD108453-1G BLDPBD108453-250MG BLDPBD108453-10G BLDPBD108453-100G BLDPBD108453-25G BLDPBD108453-5G
6-Brom-3-nitro-4-chinolinol 97%
6-Brom-3-nitro-4-chinolinol

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅BrN₂O₃
Molecular Weight: 269,0516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800413
CAS-Nummer: 853908-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR