6-Brom-[1,2,4]triazolo[4,3-a]pyridin 97%

BLDPBD108308-5GEA 192 EUR
BLDPBD108308-5G BLDPBD108308-250MG BLDPBD108308-10G BLDPBD108308-25G BLDPBD108308-1G
6-Brom-[1,2,4]triazolo[4,3-a]pyridin 97%
6-Brom-[1,2,4]triazolo[4,3-a]pyridin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₄BrN₃
Molecular Weight: 198,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689506
CAS-Nummer: 108281-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR