4,5-Difluorphthalsäureanhydrid 98%

BLDPBD10776-250MGEA 9 EUR
BLDPBD10776-0.025G BLDPBD10776-250MG BLDPBD10776-1G BLDPBD10776-25G BLDPBD10776-5G BLDPBD10776-10G BLDPBD10776-100G
4,5-Difluorphthalsäureanhydrid 98%
4,5-Difluorphthalsäureanhydrid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₂F₂O₃
Molecular Weight: 184,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08695033
CAS-Nummer: 18959-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR