3-Methoxy-o-toluidin 97%

BLDPBD10680-1GEA 13 EUR
BLDPBD10680-1G BLDPBD10680-25G BLDPBD10680-5G BLDPBD10680-100G BLDPBD10680-0.025G BLDPBD10680-10G
3-Methoxy-o-toluidin 97%
3-Methoxy-o-toluidin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₁₁NO
Molecular Weight: 137,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06412568
CAS-Nummer: 19500-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR