2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%

BLDPBD1065-500GEA 389 EUR
BLDPBD1065-500G BLDPBD1065-10G BLDPBD1065-0.025G BLDPBD1065-1G BLDPBD1065-100G BLDPBD1065-5G BLDPBD1065-25G
2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₂₁NO₂
Molecular Weight: 295,382 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134280
CAS-Nummer: 81477-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR