4-Brom-N-ethylbenzolsulfonamid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD10628-10GEA 192 EUR
BLDPBD10628-10G BLDPBD10628-1G BLDPBD10628-5G BLDPBD10628-25G
4-Brom-N-ethylbenzolsulfonamid 98%
4-Brom-N-ethylbenzolsulfonamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₀BrNO₂S
Molecular Weight: 264,1395 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01215100
CAS-Nummer: 1984-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR