Isobutyl-4-hydroxybenzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD105739-500GEA 55 EUR
BLDPBD105739-500G BLDPBD105739-100G BLDPBD105739-25G BLDPBD105739-1000G
Isobutyl-4-hydroxybenzoat 98%
Isobutyl-4-hydroxybenzoat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020167
CAS-Nummer: 4247-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR