6-Fluor-o-toluidin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD10550-1GEA 6 EUR
BLDPBD10550-1G BLDPBD10550-100G BLDPBD10550-5G BLDPBD10550-25G BLDPBD10550-10G
6-Fluor-o-toluidin 97%
6-Fluor-o-toluidin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₈FN
Molecular Weight: 125,1435 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06658252
CAS-Nummer: 443-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR