2-Fluor-5-methylphenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD10542-5GEA 23 EUR
BLDPBD10542-5G BLDPBD10542-25G BLDPBD10542-1G BLDPBD10542-10G
2-Fluor-5-methylphenylboronsäure 97%
2-Fluor-5-methylphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₈BFO₂
Molecular Weight: 153,9466 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428512
CAS-Nummer: 166328-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR