5-Methylimidazo[1,2-a]pyridin 98%

BLDPBD104867-250MGEA 33 EUR
BLDPBD104867-250MG BLDPBD104867-5G BLDPBD104867-1G
5-Methylimidazo[1,2-a]pyridin 98%
5-Methylimidazo[1,2-a]pyridin

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂
Molecular Weight: 132,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06200991
CAS-Nummer: 933-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR