2-Fluor-4-hydroxybenzoesäure 98%
|
|
BLDPBD10382-5GEA
24
EUR
BLDPBD10382-0.025G
BLDPBD10382-5G
BLDPBD10382-25G
BLDPBD10382-100G
BLDPBD10382-1G
2-Fluor-4-hydroxybenzoesäure 98%
2-Fluoro-4-hydroxybenzoic acid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₅FO₃ Molecular Weight: 156,1112 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310985 CAS-Nummer: 65145-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...