2,4-Dinitro-5-fluortoluol 95%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD10376-250MGEA
10
EUR
BLDPBD10376-250MG
2,4-Dinitro-5-fluortoluol 95%
2,4-Dinitro-5-fluortoluol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₅FN₂O₄ Molecular Weight: 200,1240 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205111 CAS-Nummer: 349-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...