6-Nitro-2-benzothiazolinone 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD10316-5GEA 108 EUR
BLDPBD10316-5G BLDPBD10316-25G BLDPBD10316-250MG BLDPBD10316-1G
6-Nitro-2-benzothiazolinone 97%
6-Nitro-2-benzothiazolinone

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄N₂O₃S
Molecular Weight: 196,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00239350
CAS-Nummer: 28620-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR