8-Amino-5-bromisochinolin 98%
|
|
BLDPBD103021-1GEA
43
EUR
BLDPBD103021-1G
BLDPBD103021-250MG
BLDPBD103021-5G
8-Amino-5-bromisochinolin 98%
8-Amino-5-bromoisoquinoline
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₉H₇BrN₂ Molecular Weight: 223,0693 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261047 CAS-Nummer: 90721-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...