Folsäure 98%
|
|
BLDPBD1030-5GEA
6
EUR
BLDPBD1030-5G
BLDPBD1030-10G
BLDPBD1030-100G
BLDPBD1030-1G
BLDPBD1030-500G
BLDPBD1030-25G
BLDPBD1030-1000G
Folsäure 98%
Folsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₉H₁₉N₇O₆ Molecular Weight: 441,41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079305 CAS-Nummer: 59-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...