5-Brom-2,1,3-benzothiadiazol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD102598-1GEA 39 EUR
BLDPBD102598-1G BLDPBD102598-10G BLDPBD102598-5G BLDPBD102598-250MG
5-Brom-2,1,3-benzothiadiazol 95%
5-Bromo-2,1,3-benzothiadiazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₃BrN₂S
Molecular Weight: 215,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00460091
CAS-Nummer: 1753-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR