5-Chlor-2-nitrodiphenylamin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD0983-10GEA 41 EUR
BLDPBD0983-10G BLDPBD0983-25G BLDPBD0983-100G BLDPBD0983-5G BLDPBD0983-1G
5-Chlor-2-nitrodiphenylamin 98%
5-Chlor-2-nitrodiphenylamin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₉ClN₂O₂
Molecular Weight: 248,6700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007287
CAS-Nummer: 25781-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR