3-(2,5-Dimethoxyphenyl)propionsäure 98%

BLDPBD0610-1GEA 25 EUR
BLDPBD0610-1G BLDPBD0610-5G BLDPBD0610-25G
3-(2,5-Dimethoxyphenyl)propionsäure 98%
3-(2,5-Dimethoxyphenyl)propionsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060325
CAS-Nummer: 10538-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR