2-(4-Bromophenyl)thiophene 97%

BLDPBD0382-5GEA 277 EUR
BLDPBD0382-5G BLDPBD0382-1G BLDPBD0382-250MG
2-(4-Bromophenyl)thiophene 97%
2-(4-Bromophenyl)thiophene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₇BrS
Molecular Weight: 239,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05664340
CAS-Nummer: 40133-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR