4-tert-Butylphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD0263-5GEA 7 EUR
BLDPBD0263-5G BLDPBD0263-10G BLDPBD0263-500G BLDPBD0263-1G BLDPBD0263-100G BLDPBD0263-1000G BLDPBD0263-25G
4-tert-Butylphenylboronsäure 98%
4-tert-Butylphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₅BO₂
Molecular Weight: 178,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01009697
CAS-Nummer: 123324-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR